accessibility menu, dialog, popup

UserName

CAS RN 54314-85-1

IUPAC Name: [3-(benzyloxy)propyl]triphenylphosphanium bromide
Synonyms: [3-(benzyloxy)propyl]triphenylphosphanium bromide
Molecular Weight (g/mol): 491.41
Molecular Formula: C28H28BrOP
InChi Key: DWYCJWXMZGVGJV-UHFFFAOYSA-M
SMILES: [Br-].C(COCC1=CC=CC=C1)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results