accessibility menu, dialog, popup

UserName

CH3NNNNH2N

CAS RN 5422-44-6

IUPAC Name: 1-methyl-1H-1,2,3,4-tetrazol-5-amine
Synonyms: 1-methyl-1,2,3,4-tetrazol-5-amine
Molecular Weight (g/mol): 99.1
Molecular Formula: C2H5N5
InChi Key: GTKOKCQMHAGFSM-UHFFFAOYSA-N
SMILES: CN1N=NN=C1N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results