accessibility menu, dialog, popup

UserName

CAS RN 5400-80-6

IUPAC Name: 2-bromo-2,3-dihydro-1H-inden-1-ol
Synonyms: 2-bromo-2,3-dihydro-1H-inden-1-ol
Molecular Weight (g/mol): 213.07
Molecular Formula: C9H9BrO
InChi Key: RTESDSDXFLYAKZ-UHFFFAOYNA-N
SMILES: OC1C(Br)CC2=CC=CC=C12
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)