CAS RN 54-88-6
IUPAC Name:
N,N,3-trimethyl-4-(2-phenyldiazen-1-yl)aniline
Synonyms:
N,N,3-trimethyl-4-(2-phenyldiazen-1-yl)aniline
Molecular Weight (g/mol):
239.32
Molecular Formula:
C15H17N3
InChi Key:
QXTOLYZIJUDOIT-UHFFFAOYSA-N
SMILES:
CN(C)C1=CC=C(N=NC2=CC=CC=C2)C(C)=C1
1
–
1
of
1
results
1
4-(Dimethylamino)-2-methylazobenzene 96.0+%, TCI America™
CAS: 54-88-6 Molecular Formula: C15H17N3 Molecular Weight (g/mol): 239.322 MDL Number: MFCD00008306 InChI Key: QXTOLYZIJUDOIT-UHFFFAOYSA-N Synonym: N,N-Dimethyl-4-phenylazo-m-toluidine PubChem CID: 5916 IUPAC Name: N,N,3-trimethyl-4-phenyldiazenylaniline SMILES: CC1=C(C=CC(=C1)N(C)C)N=NC2=CC=CC=C2