accessibility menu, dialog, popup

UserName

CAS RN 5398-77-6

IUPAC Name: 4-methanesulfonylbenzaldehyde
Synonyms: 4-methanesulfonylbenzaldehyde
Molecular Weight (g/mol): 184.21
Molecular Formula: C8H8O3S
InChi Key: PSVPUHBSBYJSMQ-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=C(C=O)C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results