accessibility menu, dialog, popup

UserName

H3CH3C(S)NHO(S)NO(S)NHO(S)CH3HNO(S)OHONHO(S)CH3HNO(S)HONHO(S)OHOHNO(S)HOONHO(S)OHOHNO(S)CH3NHOHNO(S)NH2ONHO(S)NO(S)HONHO(S)HNO(S)NH2NHO(S)HNONO(S)NH2NHNH2HNCH3H3CCH3H3COHN(S)OHOONH(S)OHO

CAS RN 53917-42-3

IUPAC Name: (4S)-4-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-{2-[(2S)-2-{[(2S)-1-[(2S)-2-[(2S)-2-[(2S)-6-amino-2-[(2S)-2-({1-[(2S)-2-amino-5-carbamimidamidopentanoyl]pyrrolidin-2-yl}formamido)-3-methylbutanamido]hexanamido]-3-methylbutanamido]-3-(4-hydroxyphenyl)propanoyl]pyrrolidin-2-yl]formamido}-3-carbamoylpropanamido]acetamido}propanamido]-4-carboxybutanamido]-3-carboxypropanamido]-4-carboxybutanamido]-3-hydroxypropanamido]propanamido]-4-{[(1S)-1-{[(2S)-1-[(2S)-2-{[(1S)-1-{[(1S)-3-carboxy-1-{[(1S)-1-carboxy-2-phenylethyl]carbamoyl}propyl]carbamoyl}-3-methylbutyl]carbamoyl}pyrrolidin-1-yl]-1-oxo-3-phenylpropan-2-yl]carbamoyl}ethyl]carbamoyl}butanoic acid
Synonyms: (4S)-4-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-{2-[(2S)-2-{[(2S)-1-[(2S)-2-[(2S)-2-[(2S)-6-amino-2-[(2S)-2-({1-[(2S)-2-amino-5-carbamimidamidopentanoyl]pyrrolidin-2-yl}formamido)-3-methylbutanamido]hexanamido]-3-methylbutanamido]-3-(4-hydroxyphenyl)propanoyl]pyrrolidin-2-yl]formamido}-3-carbamoylpropanamido]acetamido}propanamido]-4-carboxybutanamido]-3-carboxypropanamido]-4-carboxybutanamido]-3-hydroxypropanamido]propanamido]-4-{[(1S)-1-{[(2S)-1-[(2S)-2-{[(1S)-1-{[(1S)-3-carboxy-1-{[(1S)-1-carboxy-2-phenylethyl]carbamoyl}propyl]carbamoyl}-3-methylbutyl]carbamoyl}pyrrolidin-1-yl]-1-oxo-3-phenylpropan-2-yl]carbamoyl}ethyl]carbamoyl}butanoic acid
Molecular Weight (g/mol): 2465.71
Molecular Formula: C112H165N27O36
InChi Key: ZYDMZKPAPSZILB-XDGGXQJNSA-N
SMILES: CC(C)C[C@H](NC(=O)[C@@H]1CCCN1C(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@H](C)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](C)NC(=O)[C@H](CO)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](C)NC(=O)CNC(=O)[C@H](CC(N)=O)NC(=O)[C@@H]1CCCN1C(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)[C@@H](NC(=O)[C@H](CCCCN)NC(=O)[C@@H](NC(=O)C1CCCN1C(=O)[C@@H](N)CCCNC(N)=N)C(C)C)C(C)C)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results