accessibility menu, dialog, popup

UserName

CH3OHONHCl

CAS RN 53716-49-7

IUPAC Name: 2-(6-chloro-9H-carbazol-2-yl)propanoic acid
Synonyms: carprofen rimadyl carprofene carprofeno ridamyl 卡洛芬 imadyl C15H12ClNO2
Molecular Weight (g/mol): 273.72
Molecular Formula: C15H12ClNO2
InChi Key: PUXBGTOOZJQSKH-UHFFFAOYNA-N
SMILES: CC(C(O)=O)C1=CC2=C(C=C1)C1=C(N2)C=CC(Cl)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results