accessibility menu, dialog, popup

UserName

CAS RN 5367-26-0

ONOONO

CAS RN 5367-26-0

IUPAC Name: 4-[5-(4-nitrophenoxy)pentyl]morpholine
Synonyms: 4-[5-(4-nitrophenoxy)pentyl]morpholine
Molecular Weight (g/mol): 294.35
Molecular Formula: C15H22N2O4
InChi Key: QTKPBWWPUDOQGJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(OCCCCCN2CCOCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results