accessibility menu, dialog, popup

UserName

CAS RN 536-90-3

IUPAC Name: 3-methoxyaniline
Synonyms: M-anisidine m-anisidine antimonyl tartrate M-methoxyaniline
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: NCBZRJODKRCREW-UHFFFAOYSA-N
SMILES: COC1=CC=CC(N)=C1
Boiling Point 
  • (7)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (7)
  • (7)
  • (3)
  • (4)
  • (6)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (7)