accessibility menu, dialog, popup

UserName

CAS RN 536-50-5

IUPAC Name: 1-(4-methylphenyl)ethan-1-ol
Synonyms: benzenemethanol, α,4-dimethyl-
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: JESIHYIJKKUWIS-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=C(C)C=C1
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)