CAS RN 536-25-4
IUPAC Name:
methyl 3-amino-4-hydroxybenzoate
Synonyms:
orthoform orthoderm orthocaine 奥索卡因
Molecular Weight (g/mol):
167.16
Molecular Formula:
C8H9NO3
InChi Key:
VNQABZCSYCTZMS-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=C(O)C(N)=C1
1
–
1
of
1
results
1
Promotions Available
Methyl 3-amino-4-hydroxybenzoate, 95%
Methyl 3-amino-4-hydroxybenzoate, 95%
CAS: 536-25-4 Molecular Formula: C8H9NO3 Molecular Weight (g/mol): 167.164 MDL Number: MFCD00017095 InChI Key: VNQABZCSYCTZMS-UHFFFAOYSA-N Synonym: orthocaine,orthoform,aminobenz,orthoderm,3-amino-4-hydroxybenzoic acid methyl ester,benzoic acid, 3-amino-4-hydroxy-, methyl ester,methyl m-amino-p-hydroxybenzoate,methyl 4-hydroxy-3-aminobenzoate,methyl 3-amino-4-hydroxybenzenecarboxylate,unii-jk16yi13qk PubChem CID: 10815 ChEBI: CHEBI:34904 IUPAC Name: methyl 3-amino-4-hydroxybenzoate SMILES: COC(=O)C1=CC(=C(C=C1)O)N