CAS RN 53475-15-3
IUPAC Name:
3-(methylsulfanyl)butan-2-one
Synonyms:
3-(methylsulfanyl)butan-2-one
Molecular Weight (g/mol):
118.19
Molecular Formula:
C5H10OS
InChi Key:
HFVLNCDRAMUMCC-UHFFFAOYNA-N
SMILES:
CSC(C)C(C)=O
1
–
1
of
1
results
1
3-Methylthio-2-butanone 98.0+%, TCI America™
CAS: 53475-15-3 Molecular Formula: C5H10OS Molecular Weight (g/mol): 118.19 MDL Number: MFCD00008761 InChI Key: HFVLNCDRAMUMCC-UHFFFAOYNA-N Synonym: 3-methylthio-2-butanone,2-butanone, 3-methylthio,3-methylmercapto-2-butanone,3-methylsulfanyl butan-2-one,dl-3-methylthio-2-butanone,dl-3-methylthio butanone,acmc-1auk9,2-butanone,3-methylthio,3-methylsulfanyl-2-butanone PubChem CID: 103788 IUPAC Name: 3-(methylsulfanyl)butan-2-one SMILES: CSC(C)C(C)=O