accessibility menu, dialog, popup

UserName

H3CNHCH3HCl

CAS RN 5326-84-1

IUPAC Name: dipropylamine hydrochloride
Synonyms: dipropylamine hydrochloride
Molecular Weight (g/mol): 137.65
Molecular Formula: C6H16ClN
InChi Key: GAZIBGHLWYHBDT-UHFFFAOYSA-N
SMILES: Cl.CCCNCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results