accessibility menu, dialog, popup

UserName

CAS RN 53255-78-0

IUPAC Name: 3,4-dibromothieno[2,3-b]thiophene
Synonyms: 3,4-dibromothieno[2,3-b]thiophene
Molecular Weight (g/mol): 298.01
Molecular Formula: C6H2Br2S2
InChi Key: BUGQRBSPOFSLQS-UHFFFAOYSA-N
SMILES: BrC1=CSC2=C1C(Br)=CS2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results