accessibility menu, dialog, popup

UserName

CH3CH3H3COOI

CAS RN 53064-79-2

IUPAC Name: iodomethyl 2,2-dimethylpropanoate
Synonyms: iodomethyl 2,2-dimethylpropanoate
Molecular Weight (g/mol): 242.06
Molecular Formula: C6H11IO2
InChi Key: PELJISAVHGXLAL-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)OCI

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results