accessibility menu, dialog, popup

UserName

CAS RN 530-56-3

IUPAC Name: 4-(hydroxymethyl)-2,6-dimethoxyphenol
Synonyms: 4-(hydroxymethyl)-2,6-dimethoxyphenol
Molecular Weight (g/mol): 184.19
Molecular Formula: C9H12O4
InChi Key: LUOAEJWSKPQLJD-UHFFFAOYSA-N
SMILES: COC1=CC(CO)=CC(OC)=C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results