accessibility menu, dialog, popup

UserName

HClH2NN

CAS RN 530-50-7

IUPAC Name: hydrogen 1,1-diphenylhydrazine chloride
Synonyms: hydrogen 1,1-diphenylhydrazine chloride
Molecular Weight (g/mol): 220.7
Molecular Formula: C12H13ClN2
InChi Key: MIVUDWFNUOXEJM-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NN(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results