CAS RN 5293-97-0
IUPAC Name:
bis(2-chlorophenyl)methanone
Synonyms:
bis(2-chlorophenyl)methanone
Molecular Weight (g/mol):
251.11
Molecular Formula:
C13H8Cl2O
InChi Key:
DRDRZHJTTDSOPK-UHFFFAOYSA-N
SMILES:
ClC1=CC=CC=C1C(=O)C1=CC=CC=C1Cl
1
–
1
of
1
results
1
Promotions Available
2,2'-Dichlorobenzophenone, 98+%
2,2'-Dichlorobenzophenone, 98+%
CAS: 5293-97-0 Molecular Formula: C13H8Cl2O Molecular Weight (g/mol): 251.106 MDL Number: MFCD00039303 InChI Key: DRDRZHJTTDSOPK-UHFFFAOYSA-N Synonym: 2,2'-dichlorobenzophenone,bis 2-chlorophenyl methanone,methanone, bis 2-chlorophenyl,unii-5y7f5u8ant,5y7f5u8ant,benzophenone, 2,2'-dichloro,bis 2-chlorophenyl ketone,acmc-1ao0c,benzophenone,2'-dichloro,2,2-dichlorobenzophenone PubChem CID: 347097 IUPAC Name: bis(2-chlorophenyl)methanone SMILES: C1=CC=C(C(=C1)C(=O)C2=CC=CC=C2Cl)Cl