accessibility menu, dialog, popup

UserName

CH3OOOCH3O

CAS RN 529-68-0

IUPAC Name: ethyl 2-(acetyloxy)benzoate
Synonyms: ethyl 2-(acetyloxy)benzoate
Molecular Weight (g/mol): 208.21
Molecular Formula: C11H12O4
InChi Key: UYDSGXAKLVZWIJ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC=C1OC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results