accessibility menu, dialog, popup

UserName

CAS RN 5284-66-2

IUPAC Name: propane-1-sulfonic acid
Synonyms: propanesulfonic acid
Molecular Weight (g/mol): 124.15
Molecular Formula: C3H8O3S
InChi Key: KCXFHTAICRTXLI-UHFFFAOYSA-N
SMILES: CCCS(O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results