accessibility menu, dialog, popup

UserName

BrOON

CAS RN 52817-13-7

IUPAC Name: 6-bromo-4-oxo-4H-chromene-3-carbonitrile
Synonyms: 6-bromo-4-oxochromene-3-carbonitrile
Molecular Weight (g/mol): 250.05
Molecular Formula: C10H4BrNO2
InChi Key: MGVVCEKLWMZFLS-UHFFFAOYSA-N
SMILES: BrC1=CC=C2OC=C(C#N)C(=O)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results