accessibility menu, dialog, popup

UserName

OHNN

CAS RN 528-71-2

IUPAC Name: 1,5-dihydrophenazin-1-one
Synonyms: 5H-phenazin-1-one
Molecular Weight (g/mol): 196.21
Molecular Formula: C12H8N2O
InChi Key: KLJWTLFECGZQCL-UHFFFAOYSA-N
SMILES: O=C1C=CC=C2NC3=CC=CC=C3N=C12
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results