accessibility menu, dialog, popup

UserName

CAS RN 52711-30-5

IUPAC Name: 1-bromo-2-(methoxymethyl)benzene
Synonyms: 1-bromo-2-(methoxymethyl)benzene
Molecular Weight (g/mol): 201.06
Molecular Formula: C8H9BrO
InChi Key: QFAZLCRHLRJNAW-UHFFFAOYSA-N
SMILES: COCC1=CC=CC=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results