CAS RN 52085-14-0
IUPAC Name:
4-(benzyloxy)-2-hydroxybenzaldehyde
Synonyms:
4-(benzyloxy)-2-hydroxybenzaldehyde
Molecular Weight (g/mol):
228.25
Molecular Formula:
C14H12O3
InChi Key:
AMLKEDBYDOCGEG-UHFFFAOYSA-N
SMILES:
OC1=C(C=O)C=CC(OCC2=CC=CC=C2)=C1
1
–
1
of
1
results
1
Promotions Available
4-Benzyloxy-2-hydroxybenzaldehyde, 99%
4-Benzyloxy-2-hydroxybenzaldehyde, 99%
CAS: 52085-14-0 Molecular Formula: C14H12O3 Molecular Weight (g/mol): 228.247 MDL Number: MFCD01075698 InChI Key: AMLKEDBYDOCGEG-UHFFFAOYSA-N Synonym: 4-benzyloxy-2-hydroxybenzaldehyde,benzaldehyde, 2-hydroxy-4-phenylmethoxy,4-benzyloxy-2-hydroxy-benzaldehyde,2-hydroxy-4-phenylmethoxy benzaldehyde,acmc-20amov,4-benzyloxysalicylaldehyde,4-benzyloxy-2-hydoxy-benzaldehyde,4-bezyloxy-2-hydroxy-benzaldehyde,4-benzyloxy-2-hydroxy benzaldehyde PubChem CID: 561235 IUPAC Name: 2-hydroxy-4-phenylmethoxybenzaldehyde SMILES: C1=CC=C(C=C1)COC2=CC(=C(C=C2)C=O)O