CAS RN 52-76-6
CAS RN 52-76-6
IUPAC Name:
(1R,3aS,3bR,9aR,9bS,11aS)-1-ethynyl-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-ol
Synonyms:
lynestrenol lynestrenolum lynstranol exlution 17-α-ethynyloestrenol orgametril orgametrol endometril lynoestrenol ethinylestrenol
Molecular Weight (g/mol):
284.44
Molecular Formula:
C20H28O
InChi Key:
YNVGQYHLRCDXFQ-XGXHKTLJSA-N
SMILES:
C[C@]12CC[C@H]3[C@@H](CCC4=CCCC[C@H]34)[C@@H]1CC[C@@]2(O)C#C
1
–
1
of
1
results
1
Lynestrenol 98.0+%, TCI America™
CAS: 52-76-6 Molecular Formula: C20H28O Molecular Weight (g/mol): 284.443 MDL Number: MFCD00051135 InChI Key: YNVGQYHLRCDXFQ-XGXHKTLJSA-N Synonym: 19-Nor-4-pregnen-20-yn-17beta-ol PubChem CID: 5857 ChEBI: CHEBI:31790 IUPAC Name: (8R,9S,10R,13S,14S,17R)-17-ethynyl-13-methyl-2,3,6,7,8,9,10,11,12,14,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-17-ol SMILES: CC12CCC3C(C1CCC2(C#C)O)CCC4=CCCCC34