accessibility menu, dialog, popup

UserName

H3CSiH3CCH3OSiOSiH3CCH3H3COSiCH3H3CCH3OSiOSiH3CCH3CH3OSiCH3H3CCH3OSiOSiCH3H3CCH3OSiH3CCH3H3COSiCH3CH3CH3

CAS RN 51777-38-9

IUPAC Name: 2,2,6,6-tetramethyl-4,4-bis({2,2,6,6-tetramethyl-4-[(trimethylsilyl)oxy]-3,5-dioxa-2,4,6-trisilaheptan-4-yl}oxy)-3,5-dioxa-2,4,6-trisilaheptane
Synonyms: 2,2,6,6-tetramethyl-4,4-bis({2,2,6,6-tetramethyl-4-[(trimethylsilyl)oxy]-3,5-dioxa-2,4,6-trisilaheptan-4-yl}oxy)-3,5-dioxa-2,4,6-trisilaheptane
Molecular Weight (g/mol): 829.77
Molecular Formula: C24H72O10Si11
InChi Key: VLEKPQBXDHFSQO-UHFFFAOYSA-N
SMILES: C[Si](C)(C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results