accessibility menu, dialog, popup

UserName

CAS RN 51501-77-0

IUPAC Name: 2-[4,5-bis(methylsulfanyl)-2H-1,3-dithiol-2-ylidene]-4,5-bis(methylsulfanyl)-2H-1,3-dithiole
Synonyms: 2-[4,5-bis(methylsulfanyl)-1,3-dithiol-2-ylidene]-4,5-bis(methylsulfanyl)-1,3-dithiole
Molecular Weight (g/mol): 388.69
Molecular Formula: C10H12S8
InChi Key: LLLXUARUONPZIY-UHFFFAOYSA-N
SMILES: CSC1=C(SC)SC(S1)=C1SC(SC)=C(SC)S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results