accessibility menu, dialog, popup

UserName

CAS RN 5123-55-7

HOO(S)NOO

CAS RN 5123-55-7

IUPAC Name: (2S)-2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)-3-phenylpropanoic acid
Synonyms: (2S)-2-(1,3-dioxoisoindol-2-yl)-3-phenylpropanoic acid
Molecular Weight (g/mol): 295.29
Molecular Formula: C17H13NO4
InChi Key: VAYRSTHMTWUHGE-AWEZNQCLSA-N
SMILES: OC(=O)[C@H](CC1=CC=CC=C1)N1C(=O)C2=CC=CC=C2C1=O
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results