accessibility menu, dialog, popup

UserName

CAS RN 512-35-6

SOOOSOO

CAS RN 512-35-6

IUPAC Name: benzenesulfonyl benzenesulfonate
Synonyms: benzenesulfonyl benzenesulfonate
Molecular Weight (g/mol): 298.33
Molecular Formula: C12H10O5S2
InChi Key: MLWPJXZKQOPTKZ-UHFFFAOYSA-N
SMILES: O=S(=O)(OS(=O)(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results