accessibility menu, dialog, popup

UserName

CAS RN 511295-01-5

IUPAC Name: (2,3,5,6-tetrafluorophenyl)boronic acid
Synonyms: 2,3,5,6-tetrafluorophenylboronic acid
Molecular Weight (g/mol): 193.89
Molecular Formula: C6H3BF4O2
InChi Key: SORPNEUIIZJCBF-UHFFFAOYSA-N
SMILES: OB(O)C1=C(F)C(F)=CC(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results