CAS RN 50910-55-9
IUPAC Name:
2-amino-3,5-dibromobenzaldehyde
Synonyms:
2-amino-3,5-dibromobenzaldehyde
Molecular Weight (g/mol):
278.93
Molecular Formula:
C7H5Br2NO
InChi Key:
RCPAZWISSAVDEA-UHFFFAOYSA-N
SMILES:
NC1=C(Br)C=C(Br)C=C1C=O
1
–
1
of
1
results
1
2-Amino-3,5-dibromobenzaldehyde, 97%
CAS: 50910-55-9 Molecular Formula: C7H5Br2NO Molecular Weight (g/mol): 278.931 MDL Number: MFCD00671100 InChI Key: RCPAZWISSAVDEA-UHFFFAOYSA-N Synonym: 2-amino-3,5-dibromo-benzaldehyde,3,5-dibromoanthranilaldehyde,3,5-dibromo-2-aminobenzaldehyde,benzaldehyde, 2-amino-3,5-dibromo,pubchem8245,acmc-209kqx,bromhexine impurity b,ksc272a2l,2-azanyl-3,5-bis bromanyl benzaldehyde,2-amino-3,5-dibromobenzaldehyde PubChem CID: 688305 IUPAC Name: 2-amino-3,5-dibromobenzaldehyde SMILES: C1=C(C=C(C(=C1Br)N)C=O)Br