accessibility menu, dialog, popup

UserName

H3COClF

CAS RN 501-29-1

IUPAC Name: 1-chloro-2-fluoro-4-methoxybenzene
Synonyms: 1-chloro-2-fluoro-4-methoxybenzene
Molecular Weight (g/mol): 160.57
Molecular Formula: C7H6ClFO
InChi Key: FAVMSTGWFRDCRR-UHFFFAOYSA-N
SMILES: COC1=CC=C(Cl)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results