CAS RN 500199-09-7
IUPAC Name:
2-bromo-3-tetradecylthiophene
Synonyms:
2-bromo-3-tetradecylthiophene
Molecular Weight (g/mol):
359.41
Molecular Formula:
C18H31BrS
InChi Key:
GOUPSYVDGUEWMF-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCC1=C(Br)SC=C1
1
–
1
of
1
results
1
2-Bromo-3-tetradecylthiophene 95.0+%, TCI America™
CAS: 500199-09-7 Molecular Formula: C18H31BrS Molecular Weight (g/mol): 359.41 InChI Key: GOUPSYVDGUEWMF-UHFFFAOYSA-N PubChem CID: 67293590 IUPAC Name: 2-bromo-3-tetradecylthiophene SMILES: CCCCCCCCCCCCCCC1=C(SC=C1)Br