accessibility menu, dialog, popup

UserName

CH3HOOOHO

CAS RN 499-49-0

IUPAC Name: 5-methylbenzene-1,3-dicarboxylic acid
Synonyms: 5-methylbenzene-1,3-dicarboxylic acid
Molecular Weight (g/mol): 180.16
Molecular Formula: C9H8O4
InChi Key: PMZBHPUNQNKBOA-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(=C1)C(O)=O)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)