CAS RN 49843-98-3
CAS RN 49843-98-3
IUPAC Name:
6-chloro-2,3,4,9-tetrahydro-1H-carbazole-1-carboxamide
Synonyms:
6-chloro-2,3,4,9-tetrahydro-1H-carbazole-1-carboxamide
Molecular Weight (g/mol):
248.71
Molecular Formula:
C13H13ClN2O
InChi Key:
FUZYTVDVLBBXDL-UHFFFAOYNA-N
SMILES:
NC(=O)C1CCCC2=C1NC1=CC=C(Cl)C=C21
1
–
1
of
1
results
1
EX 527
CAS: 49843-98-3 Molecular Formula: C13H13ClN2O Molecular Weight (g/mol): 248.71 MDL Number: MFCD03009471 InChI Key: FUZYTVDVLBBXDL-UHFFFAOYSA-N Synonym: selisistat,sirt1 inhibitor iii,1h-carbazole-1-carboxamide, 6-chloro-2,3,4,9-tetrahydro,6-chloro-2,3,4,9-tetrahydro-1h-carbazole-1-carboxamide, racemic,selisistat inn,6-chloro-1,2,3,4,9-pentahydro-4ah-carbazolecarboxamide,ex-527 selisistat,ex 527 selisistat,d0e3lp,cambridge id 6859180 PubChem CID: 5113032 IUPAC Name: 6-chloro-2,3,4,9-tetrahydro-1H-carbazole-1-carboxamide SMILES: C1CC(C2=C(C1)C3=C(N2)C=CC(=C3)Cl)C(=O)N