accessibility menu, dialog, popup

UserName

CAS RN 4965-09-7

CH3HN

CAS RN 4965-09-7

IUPAC Name: 1-methyl-1,2,3,4-tetrahydroisoquinoline
Synonyms: 1-methyl-1,2,3,4-tetrahydroisoquinoline
Molecular Weight (g/mol): 147.22
Molecular Formula: C10H13N
InChi Key: QPILYVQSKNWRDD-UHFFFAOYNA-N
SMILES: CC1NCCC2=CC=CC=C12
Boiling Point 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results