CAS RN 495-41-0
CAS RN 495-41-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Phenyl 1-Propenyl Ketone 80.0+%, TCI America™
CAS: 495-41-0 Molecular Formula: C10H10O Molecular Weight (g/mol): 146.189 MDL Number: MFCD00059386 InChI Key: FUJZJBCWPIOHHN-QHHAFSJGSA-N Synonym: 1-Phenyl-2-buten-1-one PubChem CID: 231934 IUPAC Name: (E)-1-phenylbut-2-en-1-one SMILES: CC=CC(=O)C1=CC=CC=C1