accessibility menu, dialog, popup

UserName

H3COClNOO

CAS RN 4920-79-0

IUPAC Name: 2-chloro-1-methoxy-4-nitrobenzene
Synonyms: 2-chloro-4-nitroanisole
Molecular Weight (g/mol): 187.58
Molecular Formula: C7H6ClNO3
InChi Key: DLJPNXLHWMRQIQ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1Cl)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results