accessibility menu, dialog, popup

UserName

CAS RN 488-98-2

FClClCl

CAS RN 488-98-2

IUPAC Name: 1-fluoro-2-(trichloromethyl)benzene
Synonyms: 1-fluoro-2-(trichloromethyl)benzene
Molecular Weight (g/mol): 213.46
Molecular Formula: C7H4Cl3F
InChi Key: JYLVSNNSOWDQQX-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1C(Cl)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results