CAS RN 4861-58-9
IUPAC Name:
2-pentylthiophene
Synonyms:
2-pentylthiophene
Molecular Weight (g/mol):
154.27
Molecular Formula:
C9H14S
InChi Key:
NOYVOSGVFSEKPR-UHFFFAOYSA-N
SMILES:
CCCCCC1=CC=CS1
1
–
1
of
1
results
1
2-Pentylthiophene 98.0+%, TCI America™
CAS: 4861-58-9 Molecular Formula: C9H14S Molecular Weight (g/mol): 154.271 MDL Number: MFCD00041017 InChI Key: NOYVOSGVFSEKPR-UHFFFAOYSA-N Synonym: 2-n-pentylthiophene,thiophene, 2-pentyl,2-n-amylthiophene,1-methylbutylthiophene,unii-l79los1zkm,l79los1zkm,2-amylthiophene,2-pentyl thiophene,2-pentyl-thiophene,acmc-209kdh PubChem CID: 20995 IUPAC Name: 2-pentylthiophene SMILES: CCCCCC1=CC=CS1