accessibility menu, dialog, popup

UserName

CH3OOBOH3CH3CH3CCH3O

CAS RN 480424-70-2

IUPAC Name: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate
Synonyms: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate
Molecular Weight (g/mol): 262.11
Molecular Formula: C14H19BO4
InChi Key: KHBAJCWEQNVCSN-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results