CAS RN 474-58-8
CAS RN 474-58-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
β-Sitosterol -D-glucoside, ≥75% (HPLC), MilliporeSigma™ Supelco™
β-Sitosterol β-D-glucoside is a steroid glycoside having, β-sitosterol as the aglycone.
2
phyproof™ β-Sitosterol D-glucoside, ≥90% (HPLC), MilliporeSigma™ Supelco™
This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG