accessibility menu, dialog, popup

UserName

(R)(S)(S)(S)(R)(S)(R)(R)O(R)OH(R)OHHOHHHHMeMeMe

CAS RN 474-25-9

IUPAC Name: (4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11aR)-4,7-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
Synonyms: chenodeoxycholic acid|CDCA|chenodiol|chenix|chenocholic acid|chenodal|chenic acid|chenodesoxycholic acid|anthropodesoxycholic acid|gallodesoxycholic acid
Molecular Weight (g/mol): 392.58
Molecular Formula: C24H40O4
InChi Key: RUDATBOHQWOJDD-BSWAIDMHSA-N
SMILES: [H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)CC[C@@]1([H])[C@@]2([H])[C@H](O)C[C@]2([H])C[C@H](O)CC[C@]12C
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)