accessibility menu, dialog, popup

UserName

HO

CAS RN 4727-17-7

IUPAC Name: cyclopentadecanol
Synonyms: cyclopentadecanol
Molecular Weight (g/mol): 226.4
Molecular Formula: C15H30O
InChi Key: FFVHXGZXDRXFLQ-UHFFFAOYSA-N
SMILES: OC1CCCCCCCCCCCCCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results