accessibility menu, dialog, popup

UserName

CAS RN 47165-57-1

IUPAC Name: 3-tetradecyloxolane-2,5-dione
Synonyms: 3-tetradecyloxolane-2,5-dione
Molecular Weight (g/mol): 296.45
Molecular Formula: C18H32O3
InChi Key: BZECBEKZECEQRI-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCC1CC(=O)OC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results