accessibility menu, dialog, popup

UserName

CAS RN 457931-23-6

IUPAC Name: 4,7-bis(5-bromo-4-octylthiophen-2-yl)-2,1,3-benzothiadiazole
Synonyms: 4,7-bis(5-bromo-4-octylthiophen-2-yl)-2,1,3-benzothiadiazole
Molecular Weight (g/mol): 682.64
Molecular Formula: C30H38Br2N2S3
InChi Key: AMYLTENEOQNKGO-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=C(Br)SC(=C1)C1=CC=C(C2=CC(CCCCCCCC)=C(Br)S2)C2=NSN=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results