accessibility menu, dialog, popup

UserName

NOOOOOHOHOHOHH2OH2O

CAS RN 455303-00-1

IUPAC Name: 2-[(carboxymethyl)[(3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracen-2-yl)methyl]amino]acetic acid dihydrate
Synonyms: alizarin complexone dihydrate
Molecular Weight (g/mol): 421.36
Molecular Formula: C19H19NO10
InChi Key: LLNGUADXYFYGGJ-UHFFFAOYSA-N
SMILES: O.O.OC(=O)CN(CC(O)=O)CC1=CC2=C(C(O)=C1O)C(=O)C1=C(C=CC=C1)C2=O
Color 
  • (2)
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)