accessibility menu, dialog, popup

UserName

CAS RN 455-84-5

CH3FClO

CAS RN 455-84-5

IUPAC Name: 4-fluoro-3-methylbenzoyl chloride
Synonyms: 4-fluoro-3-methylbenzoyl chloride
Molecular Weight (g/mol): 172.58
Molecular Formula: C8H6ClFO
InChi Key: FMFLKTDRPXYUGP-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1F)C(Cl)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results