accessibility menu, dialog, popup

UserName

CAS RN 455-67-4

CH3OF

CAS RN 455-67-4

IUPAC Name: 1-(3-fluorophenyl)propan-1-one
Synonyms: 1-(3-fluorophenyl)propan-1-one
Molecular Weight (g/mol): 152.17
Molecular Formula: C9H9FO
InChi Key: RPMOHVRRKYJFSB-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC(F)=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results