accessibility menu, dialog, popup

UserName

CAS RN 452-76-6

IUPAC Name: 2,4-difluoro-1-methylbenzene
Synonyms: 2,4-difluoro-1-methylbenzene
Molecular Weight (g/mol): 128.12
Molecular Formula: C7H6F2
InChi Key: MPXDAIBTYWGBSL-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C=C1F
Boiling Point 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)